2-Amino-3-nitrophenol, 98%
2-Amino-3-nitrophenol, 98%
2-Amino-3-nitrophenol, 98%
Thermo Scientific Chemicals

2-Amino-3-nitrophenol, 98%

CAS: 603-85-0 | C6H6N2O3 | 154.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27472.06
also known as H27472-06
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
67.65
Online Exclusive
75.00
Save 7.35 (10%)
Each
Chemical Identifiers
CAS603-85-0
IUPAC Name2-amino-3-nitrophenol
Molecular FormulaC6H6N2O3
InChI KeyKUCWUAFNGCMZDB-UHFFFAOYSA-N
SMILESNC1=C(O)C=CC=C1[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red
FormPowder
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only