beta-Alanine methyl ester hydrochloride, 98%
beta-Alanine methyl ester hydrochloride, 98%
beta-Alanine methyl ester hydrochloride, 98%
beta-Alanine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals

beta-Alanine methyl ester hydrochloride, 98%

CAS: 3196-73-4 | C4H10ClNO2 | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27483.14
also known as H27483-14
Price (USD)
173.65
Online Exclusive
193.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
173.65
Online Exclusive
193.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS3196-73-4
IUPAC Namemethyl 3-aminopropanoate hydrochloride
Molecular FormulaC4H10ClNO2
InChI KeyXPGRZDJXVKFLHQ-UHFFFAOYSA-N
SMILESCl.COC(=O)CCN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystalline powder
Assay (Titration ex Chloride)≥97.5 to ≤102.5% (U.K. specification)
Assay from Supplier's CofA≥97.5% (HPLC, U.S. specification)
CommentPurchased in U.K. and U.S.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only