(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%
Thermo Scientific Chemicals

(S)-1-Cyclopropylethylamine, ChiPros™, 98%, ee 98+%

CAS: 195604-39-8 | C5H11N | 85.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27499.06
also known as H27499-06
Price (USD)
386.65
Special offer
Online exclusive
Ends: 30-Jun-2026
455.00
Save 68.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
386.65
Special offer
Online exclusive
Ends: 30-Jun-2026
455.00
Save 68.35 (15%)
Each
Chemical Identifiers
CAS195604-39-8
IUPAC Name1-cyclopropylethan-1-amine
Molecular FormulaC5H11N
InChI KeyIXCXVGWKYIDNOS-UHFFFAOYNA-N
SMILESCC(N)C1CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Refractive Index1.4245-1.4585 @ 20?C
Enantiomeric excess≥98.0%
Optical Rotation+27.5 ? 1.0? (neat)
Appearance (Color)Clear colorless to pale yellow
View more
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Cyclopropylethylamine acts as a chiral and organic building block in organic synthesis.

Solubility
Miscible with water.

Notes
Air sensitive. Incompatible with acids and strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Han, S. W.; Park, E. S.; Dong, J. Y.; Shin, J. S. Mechanism-Guided Engineering of μ-Transaminase to Accelerate Reductive Amination of Ketones. Adv. Synth. Catal. 2015, 357 (8), 1732-1740.
  2. Li, J.; Smith, D.; Krishnananthan, S.; Hartz, R. A.; Dasgupta, B.; Ahuja, V.; Schmitz, W. D.; Bronson, J. J.; Mathur, A.; Barrish, J. C.; Chen, B. C. An Efficient, Direct Bis-ortho-chlorination of 4-(Difluoromethoxy)aniline and Its Application to the Synthesis of BMS-665053, a Potent and Selective Pyrazinone-Containing Corticotropin-Releasing Factor-1 Receptor Antagonist. Org. Process Res. Dev. 2012, 16 (1), 156-159.