(S)-(-)-1-Boc-3-methylpiperazine, 98%
(S)-(-)-1-Boc-3-methylpiperazine, 98%
(S)-(-)-1-Boc-3-methylpiperazine, 98%
Thermo Scientific Chemicals

(S)-(-)-1-Boc-3-methylpiperazine, 98%

CAS: 147081-29-6 | C10H20N2O2 | 200.282 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27534.06
also known as H27534-06
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
229.65
Online Exclusive
255.00
Save 25.35 (10%)
Each
Chemical Identifiers
CAS147081-29-6
IUPAC Nametert-butyl 3-methylpiperazine-1-carboxylate
Molecular FormulaC10H20N2O2
InChI KeyFMLPQHJYUZTHQS-UHFFFAOYNA-N
SMILESCC1CN(CCN1)C(=O)OC(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point37-45?C
Optical Rotation-14.0? ?2? (c=1 in dioxane)
Assay (GC)≥97.5%
Appearance (Color)White to cream to yellow
FormFused solid or lumpy powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only