(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%
(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%
Thermo Scientific Chemicals

(S)-(+)-3-Fluoropyrrolidine hydrochloride, 97%

CAS: 136725-53-6 | C4H9ClFN | 125.571 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27536.MD
also known as H27536-MD
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.50
Save 7.85 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 30-Jun-2026
55.50
Save 7.85 (14%)
Each
Chemical Identifiers
CAS136725-53-6
IUPAC Name3-fluoropyrrolidine hydrochloride
Molecular FormulaC4H9ClFN
InChI KeyLENYOXXELREKGZ-UHFFFAOYNA-N
SMILESCl.FC1CCNC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)180.5-189.5?C
Optical Rotation8.5? ± 0.5? (C=4 in methanol)
Appearance (Color)Pale pink or pale cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only