cis-4-Aminocyclohexanecarboxylic acid, 97%
cis-4-Aminocyclohexanecarboxylic acid, 97%
cis-4-Aminocyclohexanecarboxylic acid, 97%
cis-4-Aminocyclohexanecarboxylic acid, 97%
Thermo Scientific Chemicals

cis-4-Aminocyclohexanecarboxylic acid, 97%

CAS: 3685-23-2 | C7H13NO2 | 143.186 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27567.06
also known as H27567-06
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS3685-23-2
IUPAC Name4-aminocyclohexane-1-carboxylic acid
Molecular FormulaC7H13NO2
InChI KeyDRNGLYHKYPNTEA-UHFFFAOYSA-N
SMILESNC1CCC(CC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥96.0% to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only