3-(1H-Tetrazol-5-yl)benzoic acid, 97%
3-(1H-Tetrazol-5-yl)benzoic acid, 97%
3-(1H-Tetrazol-5-yl)benzoic acid, 97%
Thermo Scientific Chemicals

3-(1H-Tetrazol-5-yl)benzoic acid, 97%

CAS: 73096-39-6 | C8H6N4O2 | 190.162 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27581.03
also known as H27581-03
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.90
Save 14.25
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
81.65
Special offer
Online exclusive
Ends: 28-Dec-2026
95.90
Save 14.25
Each
Chemical Identifiers
CAS73096-39-6
IUPAC Name3-(2H-1,2,3,4-tetrazol-5-yl)benzoic acid
Molecular FormulaC8H6N4O2
InChI KeyMIERACSHCALJOM-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC(=C1)C1=NNN=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (HPLC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only