(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%
(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%
(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%
(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%
Thermo Scientific Chemicals

(R)-(-)-1-Boc-3-hydroxypyrrolidine, 98%

CAS: 109431-87-0 | C9H17NO3 | 187.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27590.06
also known as H27590-06
Price (USD)
217.65
Special offer
Online exclusive
Ends: 31-Dec-2025
256.00
Save 38.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
217.65
Special offer
Online exclusive
Ends: 31-Dec-2025
256.00
Save 38.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS109431-87-0
IUPAC Nametert-butyl 3-hydroxypyrrolidine-1-carboxylate
Molecular FormulaC9H17NO3
InChI KeyAPCBTRDHCDOPNY-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCC(O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Proton NMRConforms to structure
Optical Rotation-21.5 ? 0.5? ( c=1.5 in chloroform)
Appearance (Color)White
Formcrystalline powder
Assay (GC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only