2-Chloro-4-fluorobenzeneboronic acid, 98%
2-Chloro-4-fluorobenzeneboronic acid, 98%
2-Chloro-4-fluorobenzeneboronic acid, 98%
Thermo Scientific Chemicals

2-Chloro-4-fluorobenzeneboronic acid, 98%

CAS: 313545-72-1 | C6H5BClFO2 | 174.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27591.14
also known as H27591-14
Price (USD)
606.65
Special offer
Online exclusive
Ends: 30-Jun-2026
714.00
Save 107.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
606.65
Special offer
Online exclusive
Ends: 30-Jun-2026
714.00
Save 107.35 (15%)
Each
Chemical Identifiers
CAS313545-72-1
IUPAC Name(2-chloro-4-fluorophenyl)boronic acid
Molecular FormulaC6H5BClFO2
InChI KeyXOFNMNLYGPKKOV-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(F)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5%
Assay (HPLC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only