Tetraethylgermanium, 99%
Tetraethylgermanium, 99%
Tetraethylgermanium, 99%
Thermo Scientific Chemicals

Tetraethylgermanium, 99%

CAS: 597-63-7 | C8H20Ge | 188.878 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27625.06
also known as H27625-06
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
113.00
Save 16.35 (14%)
Each
Chemical Identifiers
CAS597-63-7
IUPAC Nametetraethylgermane
Molecular FormulaC8H20Ge
InChI KeyQQXSEZVCKAEYQJ-UHFFFAOYSA-N
SMILESCC[Ge](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥98.5%
Refractive Index1.4415-1.4445 @ 20?C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only