alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%
alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%
alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%
Thermo Scientific Chemicals

alpha-(N,N-Dimethylamino)phenylacetonitrile, 97%

CAS: 827-36-1 | C10H12N2 | 160.22 g/mol
Have Questions?
Catalog NumberQuantity
H27631.06
also known as H27631-06
5 g
Catalog number H27631.06
also known as H27631-06
Price (USD)
32.20
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
32.20
Each
Chemical Identifiers
CAS827-36-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
FormLiquid
Assay (GC)≥96.0%
Refractive Index1.5110-1.5160 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only