cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals
cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals
cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals
cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-4-Hydroxy-D-proline, 98+%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H27633.03
also known as H27633-03
1 g
Catalog number H27633.03
also known as H27633-03
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Online Exclusive
78.80
Save 8.15 (10%)
Each
Add to cart
Chemical Identifiers
CAS2584-71-6
IUPAC Name(2R,4R)-4-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC5H9NO3
InChI KeyPMMYEEVYMWASQN-QWWZWVQMSA-N
SMILESO[C@H]1CN[C@H](C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Identification (FTIR)Conforms
Optical Rotation+59? ?1.5? (c=2 in water)
Assay (Non-aqueous acid-base Titration)≥98.0% to ≤102.0%
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only