1-Ethyl-3-methylimidazolium chloride, 98+%
1-Ethyl-3-methylimidazolium chloride, 98+%
1-Ethyl-3-methylimidazolium chloride, 98+%
Thermo Scientific Chemicals

1-Ethyl-3-methylimidazolium chloride, 98+%

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H27651.14
also known as H27651-14
Price (USD)
358.00
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
358.00
Each
Chemical Identifiers
CAS65039-09-0
IUPAC Name1-ethyl-3-methyl-1H-imidazol-3-ium chloride
Molecular FormulaC6H11ClN2
InChI KeyBMQZYMYBQZGEEY-UHFFFAOYSA-M
SMILES[Cl-].CCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or fused/lumpy solid
Assay (Titration ex Chloride)≥98.0 to ≤102.0%
Identification (FTIR)Conforms
Water Content (Karl Fischer Titration)<1.0%
Appearance (Color)White to cream to yellow to orange
Used in cellulose processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in cellulose processing.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
RUO – Research Use Only