4-Mercaptophenylacetic acid, 97%
4-Mercaptophenylacetic acid, 97%
4-Mercaptophenylacetic acid, 97%
Thermo Scientific Chemicals

4-Mercaptophenylacetic acid, 97%

CAS: 39161-84-7 | C8H8O2S | 168.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27658.03
also known as H27658-03
Price (USD)
89.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
89.00
Each
Chemical Identifiers
CAS39161-84-7
SpecificationsSpecification SheetSpecification Sheet
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
Assay (HPLC)≥96.0%
Identification (FTIR)Conforms
Appearance (Color)White to pale cream
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An useful synthetic intermediate for the preparation of novel anti inflammatory agents.

Solubility
Slightly soluble in water. Soluble in Dichloromethane, Ethanol and Methanol

Notes
Air sensitive. Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only