(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
Thermo Scientific Chemicals

(R)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

CAS: 23357-47-3 | C11H12O2 | 176.215 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H27673.06
also known as H27673-06
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
542.65
Online Exclusive
603.00
Save 60.35 (10%)
Each
Chemical Identifiers
CAS23357-47-3
IUPAC Name1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular FormulaC11H12O2
InChI KeyVDLWTJCSPSUGOA-UHFFFAOYNA-N
SMILESOC(=O)C1CCCC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)53.5-59.5°C
FormCrystals or powder or crystalline powder
Assay (Silylated GC)≥97.5%
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
View more
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a palonosetron intermediate. Tetrahydronaphthoic acid is used for the construction of sterically constrained protein turn models.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
I. Ernest.; J. Kalvoda.; G. Rihs.; M. Mutterd. Three novel mimics for the construction of sterically constrained protein turn models.Tetrahedron Lett. 1990, 31 (28), 4011-4014 .