3-Iodo-1-(phenylsulfonyl)indole, 95%
3-Iodo-1-(phenylsulfonyl)indole, 95%
3-Iodo-1-(phenylsulfonyl)indole, 95%
Thermo Scientific Chemicals

3-Iodo-1-(phenylsulfonyl)indole, 95%

CAS: 80360-14-1 | C14H10INO2S | 383.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27696.MD
also known as H27696-MD
Price (USD)
59.65
Online exclusive
66.00
Save 6.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
59.65
Online exclusive
66.00
Save 6.35
Each
Chemical Identifiers
CAS80360-14-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream
Assay (GC)≥94.0%
Melting Point (clear melt)121.0-133.0?C
FormPowder
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Takao Sakamoto; Kazutoshi Ohsawa. Palladium-catalyzed cyanation of aryl and heteroaryl iodides with copper(I) cyanide.J. Chem. Soc., Perkin Trans.. 1999, 1 (6),2323-2326 .
  2. Takao Sakamoto.; Yoshinori Kondo.; Nobuo Takazawa.; Hiroshi Yamanaka. Indolylzinc iodides by oxidative addition of active zinc to iodoindoles.Tetrahedron Lett. 1993, 34 (37),5955-5956 .