5-Bromo-1,4-benzodioxane, 95%
5-Bromo-1,4-benzodioxane, 95%
5-Bromo-1,4-benzodioxane, 95%
Thermo Scientific Chemicals

5-Bromo-1,4-benzodioxane, 95%

CAS: 58328-39-5 | C8H7BrO2 | 215.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H27709.03
also known as H27709-03
Price (USD)
206.65
Online exclusive
229.00
Save 22.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
206.65
Online exclusive
229.00
Save 22.35
Each
Chemical Identifiers
CAS58328-39-5
IUPAC Name5-bromo-2,3-dihydro-1,4-benzodioxine
Molecular FormulaC8H7BrO2
InChI KeyNGOZRIZXELGVHK-UHFFFAOYSA-N
SMILESBrC1=CC=CC2=C1OCCO2
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5925-1.5985 @ 20?C
Appearance (Color)Clear colorless to yellow
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only