Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride, 98+%
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride, 98+%
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride, 98+%
Thermo Scientific Chemicals

Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride, 98+%

CAS: 79269-71-9 | C18H26Cl2Ti
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27714.03
also known as H27714-03
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
93.65
Online Exclusive
104.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS79269-71-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to yellow-brown or orange to red
Proton NMRConforms to structure.
FormCrystals or Crystalline powder
Elemental Analysis(Carbon, C): 56.82-62.80%
Elemental Analysis(Hydrogen, H): 6.90-7.62%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only