(R)-(+)-2-Methyl-2-propanesulfinamide, 98%
(R)-(+)-2-Methyl-2-propanesulfinamide, 98%
(R)-(+)-2-Methyl-2-propanesulfinamide, 98%
Thermo Scientific Chemicals

(R)-(+)-2-Methyl-2-propanesulfinamide, 98%

CAS: 196929-78-9 | C4H11NOS | 121.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27724.06
also known as H27724-06
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
214.65
Online Exclusive
238.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS196929-78-9
IUPAC Name2-methylpropane-2-sulfinamide
Molecular FormulaC4H11NOS
InChI KeyCESUXLKAADQNTB-UHFFFAOYNA-N
SMILESCC(C)(C)S(N)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
Optical Rotation5.0 ± 1.5? (C=1 in Chloroform)
(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(+)-2-Methyl-2-propanesulfinamide is a chiral ligand, which is used in pharmaceutical compositions. Further, it is used in the preparation of beta-chloro sulfinamides in the synthesis of chiral azridines. It is involved in the preparation of organocatalyst for enantioselective reduction of imines. It serves as a reagent for synthesizing chiral amines. In addition to this, it is converted into P,N-sulfinyl imine ligands through condensation with aldehydes and ketones which undergoes iridium-catalyzed asymmetric hydrogenation of olefins.

Solubility
Soluble in chloroform, methanol, tetrahydrofuran, dichloromethane, dimethyl sulfoxide and most organic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.
RUO – Research Use Only

General References:

  1. Qian, X.; Tang, E.; Fan, J.; Zhu, K.; Zhu, J.; Shabaker, J.; Lo, E.; Malley, M.; Deshpande, R. A stereoselective synthesis of (S)-2-(((3-fluoro-4-methylphenoxy)carbonyl)(1-(4-((5-methyl-2-phenyloxazol-4-yl)methoxy)phenyl)ethyl)amino)acetic acid, a highly potent PPAR alpha/gamma dual agonist. Tetrahedron 2015, 71 (50), 9408-9414.
  2. Bender, A. M.; Griggs, N. W.; Anand, J. P.; Traynor, J. R.; Jutkiewicz, E. M.; Mosberg, H. I. Asymmetric Synthesis and in Vitro and in Vivo Activity of Tetrahydroquinolines Featuring a Diverse Set of Polar Substitutions at the 6 Position as Mixed-Efficacy mu Opioid Receptor/delta Opioid Receptor Ligands. ACS Chem. Neurosci. 2015, 6 (8), 1428-1435.