2-Chloro-6-cyanopyridine, 97%
2-Chloro-6-cyanopyridine, 97%
2-Chloro-6-cyanopyridine, 97%
Thermo Scientific Chemicals

2-Chloro-6-cyanopyridine, 97%

CAS: 33252-29-8 | C6H3ClN2 | 138.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27748.06
also known as H27748-06
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
225.65
Online Exclusive
250.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS33252-29-8
IUPAC Name6-chloropyridine-2-carbonitrile
Molecular FormulaC6H3ClN2
InChI KeyPGZHSVWXFKKCNR-UHFFFAOYSA-N
SMILESClC1=CC=CC(=N1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Melting Point (clear melt)82.0-92.0?C
Assay (GC)≥96.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only