Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)
Thermo Scientific Chemicals

Dichlorotetrakis[2-(2-pyridyl)phenyl]diiridium(III)

CAS: 92220-65-0 | C22H16ClIrN2 | 536.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
Catalog number H27752.ME
also known as H27752-ME
Price (USD)
-
Quantity:
500 mg
Request bulk or custom format
Chemical Identifiers
CAS92220-65-0
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Appearance (Color)Yellow
Assay (HPLC)≥98.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only