1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%
Thermo Scientific Chemicals

1-n-Butyl-3-methylimidazolium methyl sulfate, 99%

CAS: 401788-98-5 | C9H18N2O4S | 250.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H27754.18
also known as H27754-18
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
207.65
Online Exclusive
231.00
Save 23.35 (10%)
Each
Chemical Identifiers
CAS401788-98-5
IUPAC Name1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate
Molecular FormulaC9H18N2O4S
InChI KeyMEMNKNZDROKJHP-UHFFFAOYSA-M
SMILESCOS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to orange
FormViscous liquid
Assay from Suppliers CofA≥97.5% (NMR)
Impurity contentHalides: ≤100ppm
Water Content (Karl Fischer Titration)≤2000ppm (≤0.20%)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only