Thermo Scientific Chemicals

2-Hydroxycarbazole, 97%, Thermo Scientific Chemicals

Catalog number: H27765.14
25 g, Each
Thermo Scientific Chemicals

2-Hydroxycarbazole, 97%, Thermo Scientific Chemicals

Catalog number: H27765.14
25 g, Each
Quantity
Catalog number: H27765.14
also known as H27765-14
Price (USD)
Price: 709.00
Online price: 619.65
Your price:
Quantity
-

Chemical Identifiers

CAS
86-79-3
Appearance (Color)
White to cream to yellow to brown or grey
Identification (FTIR)
Conforms
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥96.0%

Description

Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazol has been found possible to distinguish between heparin, heparin derivatives, and other polyuronides of connective tissue. A new modification of the carbazole analysis is useful for determination of d-mannuronic acid in heteropolysaccharides. 2-Hydroxycarbazole was used in the synthesis of isochromene fused carbazol, (4aS,13bR)-2,5,5-trimethyl-3,4,4a,5,8,13b-hexahydroisochromeno[3,4-b]carbazole.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text