1,3-Dimethylimidazolium dimethyl phosphate, 98%
1,3-Dimethylimidazolium dimethyl phosphate, 98%
1,3-Dimethylimidazolium dimethyl phosphate, 98%
Thermo Scientific Chemicals

1,3-Dimethylimidazolium dimethyl phosphate, 98%

CAS: 654058-04-5 | C7H15N2O4P | 222.181 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
Catalog number H27788.18
also known as H27788-18
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
168.65
Online Exclusive
187.00
Save 18.35 (10%)
Each
Chemical Identifiers
CAS654058-04-5
IUPAC Name1,3-dimethyl-1H-imidazol-3-ium dimethyl phosphate
Molecular FormulaC7H15N2O4P
InChI KeyGSGLHYXFTXGIAQ-UHFFFAOYSA-M
SMILESCOP([O-])(=O)OC.CN1C=C[N+](C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear pale yellow to yellow
FormLiquid or viscous liquid
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Identification (FTIR)Conforms
Refractive Index1.4820-1.4860 @ 20?C
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. ionic liquid 1,3-dimethylimidazolium dimethylphosphate ([MMIM][DMP]) + water/ethanol/methanol mixtures exhibit properties which render them suitable as candidates for working pairs in industrial applications of absorption heat pumps or chillers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. ionic liquid 1,3-dimethylimidazolium dimethylphosphate ([MMIM][DMP]) + water/ethanol/methanol mixtures exhibit properties which render them suitable as candidates for working pairs in industrial applications of absorption heat pumps or chillers.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Qiang Li.; Yu-Cai He.; Mo Xian.; Gao Jun.; Xin Xu.; Jian-Ming Yang.; Liang-Zhi Li. Improving enzymatic hydrolysis of wheat straw using ionic liquid 1-ethyl-3-methyl imidazolium diethyl phosphate pretreatment. Bioresour. Technol. 2009, 100 (14),3570-3575 .
  2. Zongbao Hea.; Zongchang Zhao.;Xiaodong Zhang.; Hao Feng. Thermodynamic properties of new heat pump working pairs: 1,3-Dimethylimidazolium dimethylphosphate and water, ethanol and methanol.Fluid Phase Equilib. 2010, 298 (1),83-91 .