4-Phenyl-3-butyn-2-one, 96%
4-Phenyl-3-butyn-2-one, 96%
4-Phenyl-3-butyn-2-one, 96%
Thermo Scientific Chemicals

4-Phenyl-3-butyn-2-one, 96%

CAS: 1817-57-8 | C10H8O | 144.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27826.03
also known as H27826-03
Price (USD)
56.65
Online Exclusive
63.20
Save 6.55 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
56.65
Online Exclusive
63.20
Save 6.55 (10%)
Each
Chemical Identifiers
CAS1817-57-8
IUPAC Name4-phenylbut-3-yn-2-one
Molecular FormulaC10H8O
InChI KeyUPEUQDJSUFHFQP-UHFFFAOYSA-N
SMILESCC(=O)C#CC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5710-1.5770 @ 20?C
Assay (GC)≥95.0%
FormLiquid
Appearance (Color)Clear colorless to yellow
It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. P.Veeraraghavan Ramachandran.; Baoqing Gong.; Aleksandar V. Teodorovic'.; Herbert C. Brown. Selective reductions. 52. Efficient asymmetric reduction of α-acetylenic α'-fluoroalkyl ketones with either B-chlorodiisopinocampheylborane or B-isopinocampheyl-9-borabicyclo[3.3.1]nonane in high enantiomeric purity. The influence of fluoro groups in such reductions.Tetrahedron: Asymmetry. 1994, 5 (6),1061-1074 .
  2. M. Mark Midland.; Deborah C. McDowell.; Robert L. Hatch.; Alfonso Tramontano. Reduction of .alpha.,.beta.-acetylenic ketones with B-3-pinanyl-9-borabicyclo[3.3.1]nonane. High asymmetric induction in aliphatic systems.J. Am. Chem. Soc. 1980, 102 (2),867-869 .