4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals
4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-(4-Dimethylaminophenylazo)benzoic acid sodium salt, indicator grade, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H27830.06
also known as H27830-06
5 g
Catalog number H27830.06
also known as H27830-06
Price (USD)
197.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
197.00
Each
Chemical Identifiers
CAS845-46-5
IUPAC Namesodium 4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzoate
Molecular FormulaC15H14N3NaO2
InChI KeyOSCKRHPYZNTEIO-UHFFFAOYSA-M
SMILES[Na+].CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)Orange
Color ChangeFrom pH 1.0 (Red) to pH 3.0 (Yellow)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only