1-Hexylboronic acid pinacol ester, 98%
1-Hexylboronic acid pinacol ester, 98%
1-Hexylboronic acid pinacol ester, 98%
Thermo Scientific Chemicals

1-Hexylboronic acid pinacol ester, 98%

CAS: 86308-26-1 | C12H25BO2 | 212.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H27831.03
also known as H27831-03
Price (USD)
39.65
Online Exclusive
43.60
Save 3.95 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
39.65
Online Exclusive
43.60
Save 3.95 (9%)
Each
Chemical Identifiers
CAS86308-26-1
IUPAC Name2-hexyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC12H25BO2
InChI KeyLDPMCSWZEGKWLC-UHFFFAOYSA-N
SMILESCCCCCCB1OC(C)(C)C(C)(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4235-1.4275 @ 20?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only