tert-Butoxybis(dimethylamino)methane, tech. 90%
tert-Butoxybis(dimethylamino)methane, tech. 90%
tert-Butoxybis(dimethylamino)methane, tech. 90%
Thermo Scientific Chemicals

tert-Butoxybis(dimethylamino)methane, tech. 90%

CAS: 5815-08-7 | C9H24N2O | 176.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27861.06
also known as H27861-06
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
130.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS5815-08-7
IUPAC Name[(tert-butoxy)(dimethylazaniumyl)methyl]dimethylazanium
Molecular FormulaC9H24N2O
InChI KeyHXRAMSFGUAOAJR-UHFFFAOYSA-P
SMILESC[NH+](C)C(OC(C)(C)C)[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay (GC)≥88.0% (sum of 3 components)
Appearance (Color)Clear colorless to yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only