tert-Butoxybis(dimethylamino)methane, tech. 90%
tert-Butoxybis(dimethylamino)methane, tech. 90%
tert-Butoxybis(dimethylamino)methane, tech. 90%
Thermo Scientific Chemicals

tert-Butoxybis(dimethylamino)methane, tech. 90%

CAS: 5815-08-7 | C9H24N2O | 176.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H27861.14
also known as H27861-14
Price (USD)
362.65
Special offer
Online exclusive
Ends: 30-Jun-2026
427.00
Save 64.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
362.65
Special offer
Online exclusive
Ends: 30-Jun-2026
427.00
Save 64.35 (15%)
Each
Chemical Identifiers
CAS5815-08-7
IUPAC Name[(tert-butoxy)(dimethylazaniumyl)methyl]dimethylazanium
Molecular FormulaC9H24N2O
InChI KeyHXRAMSFGUAOAJR-UHFFFAOYSA-P
SMILESC[NH+](C)C(OC(C)(C)C)[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4200-1.4280 @ 20?C
Appearance (Color)Clear colorless to pale yellow
Identification (FTIR)Conforms
Impurity content≤ 5.0% DMF
FormLiquid
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only