(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals
(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals
(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(4-Isopropylphenoxy)acetyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H28036.06
also known as H28036-06
5 g
Catalog number H28036.06
also known as H28036-06
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
100.65
Online Exclusive
112.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS223128-33-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow or pink
FormLiquid
Identification (FTIR)Conforms
Refractive Index1.5170-1.5210 @ 20°C
Assay (GC)≥97.5%
Used as a primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a primary and secondary intermediate for pharmaceutical.

Solubility
Reacts with water.

Notes
Moisture Sensitive, store away from water/moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas.
RUO – Research Use Only
E. B. Nikolaenkova; I. A. Os’kina; V. A. Savel’eva; A. Ya. Tikhonov; V. A. Ryabinin; and A. N. Sinyakov. Synthesis of Nucleosides Containing a Photolabile 2-(2-Nitrophenyl)propoxycarbonyl Group. Russian Journal of Organic Chemistry. 2015, 51 (1), 141-144.