Ethyl alpha,4-dibromophenylacetate, 97+%
Ethyl alpha,4-dibromophenylacetate, 97+%
Ethyl alpha,4-dibromophenylacetate, 97+%
Thermo Scientific Chemicals

Ethyl alpha,4-dibromophenylacetate, 97+%

CAS: 77143-76-1 | C10H10Br2O2 | 321.996 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H28093.06
also known as H28093-06
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
134.65
Online Exclusive
149.00
Save 14.35 (10%)
Each
Chemical Identifiers
CAS77143-76-1
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.0%
Appearance (Color)Clear colorless to yellow
FormLiquid
Refractive Index1.5675-1.5725 @ 20?C
It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only
Achille Cappiello; Giorgio Famiglini; Pierangela Palma; Veronica Termopoli; Helga Trufelli. A new liquid chromatography-mass spectrometry approach for generic screening and quantitation of potential genotoxic alkylation compounds without derivatization. Journal of Chromatography A. 2012, 1255286-290.