4-Chloro-2-(trifluoromethyl)benzeneboronic acid, 97%
4-Chloro-2-(trifluoromethyl)benzeneboronic acid, 97%
4-Chloro-2-(trifluoromethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

4-Chloro-2-(trifluoromethyl)benzeneboronic acid, 97%

CAS: 313545-41-4 | C7H5BClF3O2 | 224.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H28351.09
also known as H28351-09
Price (USD)
248.65
Online Exclusive
276.00
Save 27.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
248.65
Online Exclusive
276.00
Save 27.35 (10%)
Each
Chemical Identifiers
CAS313545-41-4
IUPAC Name[4-chloro-2-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H5BClF3O2
InChI KeyYAPBOBGBYQQYHX-UHFFFAOYSA-N
SMILESOB(O)C1=C(C=C(Cl)C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0%
Assay (HPLC)≥96.0%
Melting Point (clear melt)174.0-184.0?C
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only