4-Cyano-4-phenylpiperidine hydrochloride, 97%
4-Cyano-4-phenylpiperidine hydrochloride, 97%
4-Cyano-4-phenylpiperidine hydrochloride, 97%
4-Cyano-4-phenylpiperidine hydrochloride, 97%
Thermo Scientific Chemicals

4-Cyano-4-phenylpiperidine hydrochloride, 97%

CAS: 51304-58-6 | C12H15ClN2 | 222.716 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H28414.09
also known as H28414-09
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
198.65
Online Exclusive
221.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS51304-58-6
IUPAC Name4-phenylpiperidine-4-carbonitrile hydrochloridyl
Molecular FormulaC12H14ClN2
InChI KeyRXZQAWBXAWPSJL-UHFFFAOYSA-N
SMILES[Cl].N#CC1(CCNCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Assay (Freebase GC)≥96.0%
Melting Point (clear melt)210.0-217.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only