3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%
3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%
3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester, 97%

CAS: 1073371-72-8 | C16H25BO3 | 276.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H28557.03
also known as H28557-03
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
122.65
Special offer
Online exclusive
Ends: 28-Dec-2026
144.00
Save 21.35
Each
Specifications
CAS1073371-72-8
Chemical Name or Material3-(4-Methoxyphenyl)-1-propylboronic acid pinacol ester
Formula Weight276.19
IUPAC Name2-[3-(4-methoxyphenyl)propyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
InChI KeyZJNJYHQBUIHZLN-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only