3-Benzylbenzeneboronic acid pinacol ester, 97%
3-Benzylbenzeneboronic acid pinacol ester, 97%
3-Benzylbenzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-Benzylbenzeneboronic acid pinacol ester, 97%

CAS: 1073355-05-1 | C19H23BO2 | 294.201 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28682.03
also known as H28682-03
Price (USD)
105.65
Special offer
Online exclusive
Ends: 28-Dec-2026
124.00
Save 18.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
105.65
Special offer
Online exclusive
Ends: 28-Dec-2026
124.00
Save 18.35
Each
Chemical Identifiers
CAS1073355-05-1
IUPAC Name2-(3-benzylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC19H23BO2
InChI KeyVNZSTZMLJUPNQM-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CC(CC2=CC=CC=C2)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)64.0-73.0?C
FormCrystals or powder or crystalline powder
Appearance (Color)White
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only