Furan-2,5-dicarboxylic acid, 98%
Furan-2,5-dicarboxylic acid, 98%
Furan-2,5-dicarboxylic acid, 98%
Thermo Scientific Chemicals

Furan-2,5-dicarboxylic acid, 98%

CAS: 3238-40-2 | C6H4O5 | 156.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28718.03
also known as H28718-03
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.20
Save 14.55 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 30-Jun-2026
99.20
Save 14.55 (15%)
Each
Chemical Identifiers
CAS3238-40-2
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (Silylated GC)≥97.5%
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only