3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester, 97%

CAS: 883738-27-0 | C18H29BN2O2 | 316.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28762.06
also known as H28762-06
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
320.65
Online Exclusive
356.00
Save 35.35 (10%)
Each
Chemical Identifiers
CAS883738-27-0
IUPAC Name1-methyl-4-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}piperazine
Molecular FormulaC18H29BN2O2
InChI KeyPESZAXRJFGLGTR-UHFFFAOYSA-N
SMILESCN1CCN(CC2=CC=CC(=C2)B2OC(C)(C)C(C)(C)O2)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder or liquid or viscous liquid as melt
Melting Point (clear melt)42.0-52.0?C
Appearance (Color)Pale cream to pale yellow
Assay (GC)≥96.0%
Refractive Index1.5125-1.5175 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only