2-Fluoro-4-(trifluoromethyl)benzeneboronic acid, 97%
2-Fluoro-4-(trifluoromethyl)benzeneboronic acid, 97%
2-Fluoro-4-(trifluoromethyl)benzeneboronic acid, 97%
Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethyl)benzeneboronic acid, 97%

CAS: 503309-11-3 | C7H5BF4O2 | 207.919 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28788.03
also known as H28788-03
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
99.65
Special offer
Online exclusive
Ends: 30-Jun-2026
117.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS503309-11-3
IUPAC Name[2-fluoro-4-(trifluoromethyl)phenyl]boronic acid
Molecular FormulaC7H5BF4O2
InChI KeySSFSVKVWAURAAM-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(C=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Proton NMRConforms to structure
Assay (Aqueous acid-base Titration)≥96.0%
Melting Point (clear melt)109.0-119.0?C
FormCrystals or powder or crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only