4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-2-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H28872.09
also known as H28872-09
10 g
Catalog number H28872.09
also known as H28872-09
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
402.65
Online Exclusive
447.00
Save 44.35 (10%)
Each
Chemical Identifiers
CAS160591-91-3
IUPAC Name(4-chloro-2-fluorophenyl)boronic acid
Molecular FormulaC6H5BClFO2
InChI KeyYBNDRTRLXPEWKQ-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)C=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (HPLC)≥96.0%
Assay (Aqueous acid-base Titration)≥96.0%
Proton NMRConforms to structure
Appearance (Color)White to pale cream
Used as intermediates for pharmaceutical and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as intermediates for pharmaceutical and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only