3-Phenyl-1-propylboronic acid pinacol ester, 97%
3-Phenyl-1-propylboronic acid pinacol ester, 97%
3-Phenyl-1-propylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

3-Phenyl-1-propylboronic acid pinacol ester, 97%

CAS: 329685-40-7 | C15H23BO2 | 246.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H28985.03
also known as H28985-03
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
329.65
Online Exclusive
366.00
Save 36.35 (10%)
Each
Chemical Identifiers
CAS329685-40-7
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.4830-1.4880 @ 20?C
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only