Potassium 3-iodophenyltrifluoroborate, 96%, Thermo Scientific Chemicals
Potassium 3-iodophenyltrifluoroborate, 96%, Thermo Scientific Chemicals
Potassium 3-iodophenyltrifluoroborate, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium 3-iodophenyltrifluoroborate, 96%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H29009.06
also known as H29009-06
5 g
Catalog number H29009.06
also known as H29009-06
Price (USD)
97.65
Online exclusive
108.00
Save 10.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
97.65
Online exclusive
108.00
Save 10.35
Each
Specifications
CAS1189097-36-6
Chemical Name or MaterialPotassium 3-iodophenyltrifluoroborate
Formula Weight309.9
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only