5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals
5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals
5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals
5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H29217.03
also known as H29217-03
1 g
Catalog number H29217.03
also known as H29217-03
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
187.65
Online Exclusive
208.00
Save 20.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS40432-84-6
IUPAC Name2-(5-bromo-1H-indol-3-yl)acetate
Molecular FormulaC10H7BrNO2
InChI KeyWTFGHMZUJMRWBK-UHFFFAOYSA-M
SMILES[O-]C(=O)CC1=CNC2=CC=C(Br)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to orange
FormCrystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only