2,6-Dichlorobenzylamine, 97+%
2,6-Dichlorobenzylamine, 97+%
2,6-Dichlorobenzylamine, 97+%
2,6-Dichlorobenzylamine, 97+%
Thermo Scientific Chemicals

2,6-Dichlorobenzylamine, 97+%

CAS: 6575-27-5 | C7H7Cl2N | 176.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Catalog number H29274.14
also known as H29274-14
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
206.00
Save 30.35 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)
175.65
Special offer
Online exclusive
Ends: 30-Jun-2026
206.00
Save 30.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS6575-27-5
IUPAC Name1-(2,6-dichlorophenyl)methanamine
Molecular FormulaC7H7Cl2N
InChI KeyVLVLNNQMURDGPM-UHFFFAOYSA-N
SMILESNCC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0%
FormLiquid
Refractive Index1.5745-1.5795 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only