Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water
Thermo Scientific Chemicals

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, may cont. 1-5% water

Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate, 97%, C32H12BF24Na, CAS Number-79060-88-1 | CAS: 79060-88-1 | C32H12BF24Na | 886.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H30014.06
Price (USD)
536.65
Special offer
Online exclusive
Ends: 30-Jun-2026
631.00
Save 94.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
536.65
Special offer
Online exclusive
Ends: 30-Jun-2026
631.00
Save 94.35 (15%)
Each
Chemical Identifiers
CAS79060-88-1
IUPAC Namesodium tetrakis[3,5-bis(trifluoromethyl)phenyl]boranuide
Molecular FormulaC32H12BF24Na
InChI KeyLTGMONZOZHXAHO-UHFFFAOYSA-N
SMILES[Na+].FC(F)(F)C1=CC(=CC(=C1)[B-](C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)(C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
Assay (special)≥96.0 to ≤104.0% based on elemental C (dry wt. basis)
Water Content (Karl Fischer Titration)≤5%
Proton NMRConforms to structure
FormPowder
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium tetrakis[3,5-bis(trifluoromethyl)phenyl]borate is a lipophilic additive of tetraphenylborate and it is used for solvent extraction of cations and solvent polymeric membrane electrodes and optodes. It is also used as an anionic phase-transfer catalyst.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. López, L. A.; López, E. Recent advances in transition metal-catalyzed C-H bond functionalization of ferrocene derivatives. Dalton Trans. 2015, 44 (22), 10128-10135.
  2. Li, M.; Chen, C. Polymerization of disubstituted acetylenes by monodentate NHC-Pd catalysts. Polym. Chem. 2015, 6 (40), 7127-7132.