4-Benzyloxy-1-bromo-2-fluorobenzene, 98%
4-Benzyloxy-1-bromo-2-fluorobenzene, 98%
4-Benzyloxy-1-bromo-2-fluorobenzene, 98%
4-Benzyloxy-1-bromo-2-fluorobenzene, 98%
Thermo Scientific Chemicals

4-Benzyloxy-1-bromo-2-fluorobenzene, 98%

CAS: 185346-79-6 | C13H10BrFO | 281.124 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H30053.06
also known as H30053-06
Price (USD)
70.65
Special offer
Online exclusive
Ends: 31-Dec-2025
82.80
Save 12.15 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 31-Dec-2025
82.80
Save 12.15 (15%)
Each
Add to cart
Chemical Identifiers
CAS185346-79-6
IUPAC Name4-(benzyloxy)-1-bromo-2-fluorobenzene
Molecular FormulaC13H10BrFO
InChI KeyBYTJTXKQBSNGCL-UHFFFAOYSA-N
SMILESFC1=C(Br)C=CC(OCC2=CC=CC=C2)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormFused solid
Melting Point (clear melt)31.0-37.0?C
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only