1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%
1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%
1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%
Thermo Scientific Chemicals

1-(2-Phenylethyl)-4-(phenylethynyl)benzene, 97%

CAS: 906650-60-0 | C22H18 | 282.386 g/mol
Have Questions?
Catalog NumberQuantity
H30058.03
also known as H30058-03
1 g
Catalog number H30058.03
also known as H30058-03
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
106.65
Online Exclusive
118.00
Save 11.35 (10%)
Each
Chemical Identifiers
CAS906650-60-0
IUPAC Name1-(2-phenylethyl)-4-(2-phenylethynyl)benzene
Molecular FormulaC22H18
InChI KeyGXNVEXGZHSEHNG-UHFFFAOYSA-N
SMILESC(CC1=CC=C(C=C1)C#CC1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream to cream to pale yellow
Assay (GC)≥96.0%
FormPowder
Melting Point (clear melt)101.0-111.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only