Methyl 3-bromo-4-hydroxybenzoate, 98%
Methyl 3-bromo-4-hydroxybenzoate, 98%
Methyl 3-bromo-4-hydroxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-bromo-4-hydroxybenzoate, 98%

CAS: 29415-97-2 | C8H7BrO3 | 231.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number H30222.09
also known as H30222-09
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
54.65
Online Exclusive
60.70
Save 6.05 (10%)
Each
Chemical Identifiers
CAS29415-97-2
IUPAC Namemethyl 3-bromo-4-hydroxybenzoate
Molecular FormulaC8H7BrO3
InChI KeyRKUNSPWAQIUGEZ-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(Br)=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)104.0-110.0?C
Assay (GC)≥97.5%
FormPowder
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A reactant used in the preparation of selective inhibitors. In unsymmetrical Ullman reaction between methyl 5bromovanillate (II) and methyl 3-bromo-4-hydroxybenzoate (III) yielded all the three possible dicarbomethoxy-dibenzo-pdioxms which were separated by repeated chromatography over silica gel. A phosphorus- and bromine-containing bifunctional monomer has been prepared from bis(chloromethyl)methylphosphine oxide and the sodium salt of methyl 3-bromo-4-hydroxybenzoate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. H. Sivriev ,; G. Borissov ,; T.E. Kron,; E.N. Tsvetkov,; M.I. Kabachnik. Polyesters based on bromine-containing bis(p-methoxycarbonylphenoxymethyl)-methylphosphine oxide. European Polymer Journal. 15 (8),771-774.
  2. B. Anjaneyulu,; T.R. Govindachari,; N. Viswanathan.Total synthesis of dl-O-methyltiliacorine . Tetrahedron. 1971, 27 (2) , 439-443.