2-Bromonicotinic acid, 97%
2-Bromonicotinic acid, 97%
2-Bromonicotinic acid, 97%
Thermo Scientific Chemicals

2-Bromonicotinic acid, 97%

CAS: 35905-85-2 | C6H4BrNO2 | 202.007 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H30239.06
also known as H30239-06
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS35905-85-2
IUPAC Name2-bromopyridine-3-carboxylic acid
Molecular FormulaC6H4BrNO2
InChI KeyMMWNKXIFVYQOTK-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CN=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Silylated GC)≥96.0%
Appearance (Color)White to cream
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only