1,4-Bis(phenylethynyl)benzene, 97%
1,4-Bis(phenylethynyl)benzene, 97%
1,4-Bis(phenylethynyl)benzene, 97%
1,4-Bis(phenylethynyl)benzene, 97%
Thermo Scientific Chemicals

1,4-Bis(phenylethynyl)benzene, 97%

CAS: 1849-27-0 | C22H14 | 278.354 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H30395.06
also known as H30395-06
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
127.65
Online Exclusive
142.00
Save 14.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS1849-27-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow
FormCrystalline powder or powder
Assay (GC)≥96.0%
Melting Point (clear melt)176-184°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only