2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%
2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%
2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%
Thermo Scientific Chemicals

2-(3-Chlorophenyl)ethylboronic acid pinacol ester, 97%

CAS: 1315277-53-2 | C14H20BClO2 | 266.572 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H30464.06
also known as H30464-06
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
109.65
Online Exclusive
122.00
Save 12.35 (10%)
Each
Chemical Identifiers
CAS1315277-53-2
IUPAC Name2-[2-(3-chlorophenyl)ethyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular FormulaC14H20BClO2
InChI KeyWZAJMSPJXKZSTL-UHFFFAOYSA-N
SMILESCC1(C)OB(CCC2=CC(Cl)=CC=C2)OC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4940-1.4990 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only